missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLT4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5109730
Additional Details : Weight : 0.01000kg
Description
Sequence of this protein is as follows: SQVSTMALHI AQADAEDSPP SLQRHSLAAR YYNWVSFPGC LARGAETRGS SRMKTFEEFP MTPTTYKGSV DNQTDSGMVL ASEEFEQIES RHRQESGFSC K
FLT4 Polyclonal antibody specifically detects FLT4 in Human samples. It is validated for Western Blot.Specifications
FLT4 | |
Polyclonal | |
Unconjugated | |
FLT4 | |
AI323512; Chy; chylous ascites; Flt4; FLT-4; FLT41; fms related tyrosine kinase 4; FMS-like tyrosine kinase 4; fms-related tyrosine kinase 4; LMPH1A; PCL; receptor protein tyrosine kinase; soluble VEGFR 3; sVEGFR 3; tyrosine-protein kinase receptor FLT4; Vascular endothelial growth factor receptor 3; vascular endothelial growth factor receptor-3; VEGF R3; VEGFR3; VEGFR-3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2324 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P35916 | |
FLT4 | |
A synthetic peptide corresponding to a sequence within amino acids 1230-1329 of human VEGFR3/FLT4 (NP_8915552) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |