missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FLRT3 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | FLRT3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
FLRT3 Polyclonal specifically detects FLRT3 in Human samples. It is validated for Western Blot.Specifications
FLRT3 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
fibronectin leucine rich transmembrane protein 3, Fibronectin-like domain-containing leucine-rich transmembrane protein 3, HH21, KIAA1469, leucine-rich repeat transmembrane protein FLRT3 | |
FLRT3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9NZU0 | |
23767 | |
Mouse myeloma cell line NS0-derived recombinant human FLRT3, Lys29-Pro528, Accession # Q9NZU0 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human FLRT3 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human FLRT1 and rhFLRT2 is observed. |
For Research Use Only