Learn More
Florisil™, for Chromatography, about 60-100 U.S. Mesh, Fisher Chemical
Brand: Fisher Chemical F/1173/50
76.70 GBP valid until 2022-10-02
Use promo code "18216" to get your promotional price.
Code : 21
Additional Details : CAS Number : 1343-88-0 Weight : 0.25000kg
Specifications
Plastic Powder Jar | |
250 g |