Learn More
Labconco™ Flexi-Bin Cart
The Labconco Flexi-Bin Cart provides for safe transport of glassware and chemical containers.
Brand: Labconco™ 09380-02
Code : NEW
Additional Details : Weight : 23.00000kg
Description
For safe transport of glassware and chemical containers
- Removable, corrosion-resistant polyethylene bin; choice of 4- or 8 in. depth (10 or 20cm); bins are interchangeable
- Supplied with three long and four short polyethylene dividers to partition bin and hold items securely
- Glacier-white, powder-coated, 18-gauge welded tubular steel frame with rubber handle grips
- Open bottom shelf for additional storage
- Four 4 in. dia. (10cm) casters (two locking)
- Fully assembled
- Lifetime guarantee
Specifications
Flexi-bin Cart | |
Safe transport of Glassware and chemical containers | |
92.4 cm | |
34.88 in. | |
400 lb. | |
Steel | |
20 kg | |
19 in. |
34.88 x 19 x 36.38 in. (88.6 x 48.3 x 92.4 cm) | |
36.38 in. | |
18G Welded 1 in. white tubular Steel frame, Rubber handle grips, removable 8 in. D Polyethylene bin, three long (20.25 in.) and four short (15.25 in.) Polyethylene dividers, 4 in. dia. casters, two with toe locks, ID tag | |
88.6 cm | |
181 kg | |
2 | |
44.09 lb. | |
48.3 cm |