missing translation for 'onlineSavingsMsg'
Learn More

Flecainide acetate, Tocris Bioscience™

Cardiac Na+ channel blocker. Antiarrhythmic

Brand:  Tocris (Bio-Techne) 147010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13469553

  • £83.79 / 10mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Open Na+ channel blocker that inhibits fast Na+ current in cardiac muscle in a use- and concentration-dependent manner. Orally-active class Ic antiarrhythmic agent.
Specifications

Chemical Identifiers

54143-56-5
474.4
flecainide acetate, flecainide monoacetate, flecainide acetate salt, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, n-piperidin-2-ylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, tambocor tn, 2,5-bis-2,2,2-trifluoroethoxy-n-2-piperidinylmethyl benzamide acetate, n-2-piperidylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, benzamide, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy-, monoacetate
CHEBI:5091
CC(=O)O.C1CCNC(C1)CNC(=O)C2=C(C=CC(=C2)OCC(F)(F)F)OCC(F)(F)F
C19H24F6N2O5
RKXNZRPQSOPPRN-UHFFFAOYSA-N
41022
acetic acid;N-(piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide

Specifications

10mg
54143-56-5
flecainide acetate, flecainide monoacetate, flecainide acetate salt, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, n-piperidin-2-ylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, tambocor tn, 2,5-bis-2,2,2-trifluoroethoxy-n-2-piperidinylmethyl benzamide acetate, n-2-piperidylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, benzamide, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy-, monoacetate
CC(=O)O.C1CCNC(C1)CNC(=O)C2=C(C=CC(=C2)OCC(F)(F)F)OCC(F)(F)F
474.4
CHEBI:5091
>99%
Flecainide acetate
C19H24F6N2O5
RKXNZRPQSOPPRN-UHFFFAOYSA-N
acetic acid;N-(piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide
41022
474.4
Videos
Documents