Learn More
Flecainide acetate, Tocris Bioscience™
Cardiac Na+ channel blocker. Antiarrhythmic
POA
Description
Description
Open Na+ channel blocker that inhibits fast Na+ current in cardiac muscle in a use- and concentration-dependent manner. Orally-active class Ic antiarrhythmic agent.