Learn More
Heidolph™ Flat Rubber Mat for Hei-SHAKE Orbital Core 2000
Product Code. 30274406 Shop All Heidolph Instruments Products
Change view
Click to view available options
For Use With (Equipment):
Hei-SHAKE Orbital Core 2000
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
30274406 Hei-SHAKE Orbital Core 2000 Each
1 options
Product Code. 30274406 Supplier Heidolph™ Supplier No. 2303080266
Only null left
Add to basket
Add to basket
Non-slip, flat rubber mat to be placed on the integrated shaker's platform.
Non-slip, flat rubber mat to be placed on the integrated shaker's platform and optionally on the perforated platform 2000, e.g. for the multi-tier attachment 2000.
Material: EPDM
Characteristics:
  • High resistance to temperatures up to 134°C
  • High resistance to acids and alkalis
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Non-Slip Mat
Material EPDM
For Use With (Equipment) Hei-SHAKE Orbital Core 2000