missing translation for 'onlineSavingsMsg'
Learn More

Manutan Flammable material cabinet, yellow, 700mm x 915mm x 459mm

Brand:  Manutan 1955R035N

Flammable material cabinet, yellow, 700mm x 915mm x 459mm

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12816919

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.