Learn More
Integra Biosciences™ Flameboy Plus Bunsen Burner
Used with common gas types such as butane/propane and natural gas.   Integra Biosciences™ Flameboy Plus Bunsen Burner has a smooth, chrome plated metal housing which is easy to clean and both UV and solvent resistant.
Brand:  Integra Biosciences™ 144000
Code : 04
Additional Details : Weight : 0.80000kg
Product Code. 11833560
  • £505.00 / Each
In Stock 5
Add to basket
Description
Description
The smooth, chrome plated metal housing is easy to clean and both UV and solvent resistant. With flame monitor and overheating protection. Functional and robust design. Convenient handling. Safe operation. FIREBOY eco
  • Ignition by a footswitch (included) or push button
  • Two adjustment knobs for air and gas allow easy fine-tuning of flame FIREBOY plus
  • Ignition by push button, or hands free operation with movement sensor or optional footswitch (not included)
  • User defined maximal burning times
  • Optional rechargeable battery
  • Portable Compatible with common gas types such as butane/propane and natural gas; can be connected to gas distribution system or gas cartridges.
Specifications
Specifications
1 Each
Natural Gas/Butane/Propane
Bunsen Burner
Videos
Documents
View accessory products