missing translation for 'onlineSavingsMsg'
Learn More

Integra Biosciences™ Flameboy™ Sterilizing Pistol

Use this handy and portable flame sterilizer with piezo-electric ignition for safe flame sterilization. Integra Biosciences™ Flameboy™ Sterilizing Pistol operates independently of any gas distribution system or battery.

Brand:  Integra Biosciences™ 145000

Code : 04

Additional Details : Weight : 0.25000kg

Product Code. 10038391

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Ideal for production plant, fermenter, laboratory or outdoor applications
  • Gas cartridge adapters are easily mounted via an intelligent “click and go” mechanism
  • Can be connected to a gas distribution system for use in a permanent installation, such as hoods and laboratory environments
  • Piezoelectric ignition
  • Ergonomic design
  • The handy pistol grip of FLAMEBOY provides convenient holding and use
  • Various gas adapters for gas connections and cartridges
  • Compatible with different gas types
In addition to gas cartridge adapters, the FLAMEBOY may also be connected to a gas distribution system or gas cylinder and used as a fixed installation in the laboratory.
Specifications

Specifications

Sterilizing Pistol
14 in.
200W x 400D x 200H mm
200 mm
230°F
1 Ea.
400 mm
Butane, Propane
200 mm
10 in.
(3) Nozzle, Adapter, Threaded Adapter
1350°C (Propane/Butane), 1300°C (Natural gas)
0.035 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers