Learn More
FK 3311, Tocris Bioscience™
Cyclooxygenase-2 (COX-2) inhibitor
Brand:  Tocris (Bio-Techne) 277610
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13440574
  • £163.17 / 10mg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Cyclooxygenase-2 (COX-2) inhibitor. Inhibits zymosan-induced prostaglandin E2 production by rat peritoneal neutrophils in vitro and adjuvant-induced arthritis in vivo. Anti-inflammatory.
Specifications
Chemical Identifiers
116686-15-8
341.329
cox-2 inhibitor v, fk3311, n-4-acetyl-2-2,4-difluorophenoxy phenyl methanesulfonamide, 4'-acetyl-2'-2,4-difluorophenoxy methanesulfonanilide, methanesulfonamide, n-4-acetyl-2-2,4-difluorophenoxy phenyl, d0z9yw, diiylgznzgpxrr-uhfffaoysa
N-[4-acetyl-2-(2,4-difluorophenoxy)phenyl]methanesulfonamide
C15H13F2NO4S
DIIYLGZNZGPXRR-UHFFFAOYSA-N
164009
CC(=O)C1=CC(=C(C=C1)NS(=O)(=O)C)OC2=C(C=C(C=C2)F)F
Specifications
10mg
116686-15-8
cox-2 inhibitor v, fk3311, n-4-acetyl-2-2,4-difluorophenoxy phenyl methanesulfonamide, 4'-acetyl-2'-2,4-difluorophenoxy methanesulfonanilide, methanesulfonamide, n-4-acetyl-2-2,4-difluorophenoxy phenyl, d0z9yw, diiylgznzgpxrr-uhfffaoysa
CC(=O)C1=CC(=C(C=C1)NS(=O)(=O)C)OC2=C(C=C(C=C2)F)F
341.329
341.33
FK 3311
C15H13F2NO4S
DIIYLGZNZGPXRR-UHFFFAOYSA-N
N-[4-acetyl-2-(2,4-difluorophenoxy)phenyl]methanesulfonamide
164009
>98%