Learn More
Eppendorf™ Fixed Angle QuickLock™ Rotor with Aerosol-tight Lid
For use with Centrifuge 5430 and Centrifuge 5430 R
Brand: Eppendorf™ 5427753001
Code : NEW
Additional Details : Weight : 1.80000kg
Description
- Aerosol-tight Eppendorf QuickLock rotors close with only 1/4 of a turn
- Saves time during every day use
- Takes repetitive stress off your wrist
Specifications
Model F-45-48-11 | |
30 x 1.5/2 mL | |
14,000 rpm | |
Fixed | |
30 |
Fixed Angle Rotor | |
20,800 x g | |
5430/5430R Centrifuges | |
Aerosol rotor lid |