missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ FITC-CHIPOpt
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
FITC-CHIPOpt is a fluorescent CHIP/STUB1 E3 ligase inhibitor. It binds to CHIP and can be used in CHIP fluorescent polarization (FP) binding assays. Enables CHIP FP binding assays to be developed for new inhibitor or CHIP ligand synthesis. FITC-CHIPOpt is the fluorescent version of CHIPOp (Cat. No. 8044).
Specifications
Specifications
| Purity | 0.95 |
| Content And Storage | Store at -20°C |
| Molecular Weight (g/mol) | 1218.35 |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction