missing translation for 'onlineSavingsMsg'
Learn More

FIT, Tocris Bioscience™

Irreversible δ agonist

Brand:  RD Systems (Bio-Techne) 1480/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13429343

  • £742.35 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective and irreversible δ-opioid agonist (EC50 = 8 nM); acts by alkylating the receptor.
Specifications

Chemical Identifiers

85951-63-9
393.549
fentanyl isothiocyanate, fit, fentanyl ncs, n-1-2-4-isothiocyanatophenyl ethyl piperidin-4-yl-n-phenylpropanamide, fit alkylating agent, tocris-1480, n-1-2-4-isothiocyanatophenyl ethyl-4-piperidinyl-n-phenylpropanamide, n-1-2-4-isothiocyanato-phenyl-ethyl-piperidin-4-yl-n-phenyl-propionamide, propanamide, n-1-2-4-isothiocyanatophenyl ethyl-4-piperidinyl-n-phenyl
N-[1-[2-(4-isothiocyanatophenyl)ethyl]piperidin-4-yl]-N-phenylpropanamide
C23H27N3OS
VDKBIFPJULZUPU-UHFFFAOYSA-N
84008
CCC(=O)N(C1CCN(CC1)CCC2=CC=C(C=C2)N=C=S)C3=CC=CC=C3

Specifications

FIT
85951-63-9
fentanyl isothiocyanate, fit, fentanyl ncs, n-1-2-4-isothiocyanatophenyl ethyl piperidin-4-yl-n-phenylpropanamide, fit alkylating agent, tocris-1480, n-1-2-4-isothiocyanatophenyl ethyl-4-piperidinyl-n-phenylpropanamide, n-1-2-4-isothiocyanato-phenyl-ethyl-piperidin-4-yl-n-phenyl-propionamide, propanamide, n-1-2-4-isothiocyanatophenyl ethyl-4-piperidinyl-n-phenyl
CCC(=O)N(C1CCN(CC1)CCC2=CC=C(C=C2)N=C=S)C3=CC=CC=C3
393.549
393.55
50mg
C23H27N3OS
VDKBIFPJULZUPU-UHFFFAOYSA-N
N-[1-[2-(4-isothiocyanatophenyl)ethyl]piperidin-4-yl]-N-phenylpropanamide
84008
>99%
Videos
Documents