missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Glass Beaker with Cover

The Fisherbrand™ Glass Beaker with Cover is an ideal accessory for the Fisherbrand™ 11211 ultrasonic cleaner. This is a safe and economical choice for any laboratory.

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 12652525

  • £14.50 / Each
Estimated Shipment: 03-06-2024
to see stock.
Add to basket

Description

Description

  • Clear glass for easy viewing
  • Stable design prevents tipping
  • Easy to open and close lid
  • Graduated design

Specifications

Specifications

Beaker
20.28 oz.
Lid
0.6 L
FB11211 Ultrasonic Cleaning Unit
Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers