missing translation for 'onlineSavingsMsg'
Learn More

Fisher Scientific™ Traceable™ One-Shot™ Conductivity Calibration Standards Certified Reference Material Producer (CRM)

Ideal for lab or field applications

Brand:  Fisherbrand

Code : 46

Additional Details : CAS Number : 71-23-8 Weight : 0.60000kg Transport : UN number : 1274 Chem class : 1 Pack group : III

 View more versions of this product

Product Code. 11744246

  • £32.79 / Pack of 6


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Single-use conductivity standards calibrate all conductivity meters and probes for maximum accuracy. Eliminate concern about external container contamination. Calibration is made in the standard vial.

Calibration within container — zero contamination

  • Traceable to NIST (National Institute of Standards and Technology)
  • Accuracy: certificate stated uncertainty or ±0.25 microsiemen or ±0.25%
  • 100% compatible with all conductivity meters
  • A2LA accreditations ISO 34 and ISO 17025 plus ISO 9001
  • Includes an individual temperature chart and step-by-step calibration instructions
  • Units labeled (micromhos/microsiemens, ohms, parts per million/TDS)
  • Single use Eliminates external contamination concerns Each standard contains 100mL

Assortment (09-328-20) contains one each of 09-328-6, -7, -8, -9, -10, and -12

Specifications

Chemical Identifiers

6/Pk.

Specifications

6/Pk.
Conductivity Calibration Standard Assortment
Videos
Documentation