missing translation for 'onlineSavingsMsg'
Learn More

Fisher Scientific™ Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM)

Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.

Brand:  Fisherbrand

Code : 46

Additional Details : CAS Number : 71-23-8 Weight : 0.47300kg Transport : UN number : 1274 Chem class : 1 Pack group : III

Product Type NIST batch tested; Resistivity: 0.2megohm
Conductivity 5μS

 View more versions of this product

Product Code. 11724236

  • £83.75 / Each


In Stock 11
Add to basket

Description

Description

Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing. Also includes step-by step calibration instructions and temperature compensation chart. Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater.
Specifications

Specifications

5μS
NIST batch tested; Resistivity: 0.2megohm
Videos
Documentation

Technical Documents