missing translation for 'onlineSavingsMsg'
Learn More

Fisher Scientific™ Fisher Scientific™ Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM)

Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.

Brand:  Fisher Scientific™ 4274

Code : 46

Additional Details : CAS Number : 71-23-8 Weight : 0.47300kg Transport : UN number : 1274 Chem class : 1 Pack group : III

Product Type NIST batch tested; Resistivity: 1.0megohm
Conductivity 1μS

 View more versions of this product

Product Code. 11734236

  • £84.20 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing. Also includes step-by step calibration instructions and temperature compensation chart. Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater.
Specifications

Specifications

1μS
NIST batch tested; Resistivity: 1.0megohm
Videos
Documentation

Technical Documents