Learn More
Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
Brand: Fisherbrand
Code : A0
Additional Details : Weight : 6.50000kg
Description
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing. Also includes step-by step calibration instructions and temperature compensation chart. Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater.Order Info
473mL (16 oz.) bottle
Specifications
NIST Individual bottle tested; Resistivity: 0.0001 mΩ | |
Calibration Solution | |
Yes | |
A2LA Conductivity Standards | |
ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material |
1 Ea. | |
Clear | |
Conductivity Standard | |
10,000 μS | |
Liquid |