missing translation for 'onlineSavingsMsg'
Learn More

Fisher Scientific™ Fisher Scientific™ Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM)

Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.

Brand:  Fisher Scientific™ 4067

Code : 46

Additional Details : Weight : 1.00000kg

Product Type NIST batch tested; Resistivity: 0.001megohm
Conductivity 1000μS

 View more versions of this product

Product Code. 11764226

  • £31.85 / Each


Estimated Shipment
20-02-2018
Add to basket

Description

Description

Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing. Also includes step-by step calibration instructions and temperature compensation chart. Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater.
Specifications

Specifications

16 oz. (475mL)
1000μS
NIST batch tested; Resistivity: 0.001megohm
Videos
Documentation

Technical Documents