Learn More
Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips
Choose from a comprehensive line of pipet tips for demanding liquid-handling applications
Brand:  Fisherbrand™
Code : 50
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12922571
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Universal fit for research grade pipetters.
  • Engineered for increased accuracy and are compatible with single-channel, multichannel, and electronic pipetters
  • DNase-/RNase-free; sterile tips are also pyrogen-free
  • Sterile and clear color
  • Filter eliminates aerosol cross-contamination without sealing or trapping samples
  • Tips allow for noncontaminated sample recovery
  • Not autoclavable
Specifications
Specifications
Universal Fit Pipettor
MicroPoint
Filtered
Clear
No
Universal Fit Pipettor
Hinged Rack
30 to 300μL
Sterile
10, 50, 100, 200μL
5.1 cm