missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Drosophila Bottles

Wide base/narrow top design prevents fly meal from falling out when bottle is turned upside down

Brand:  Fisherbrand™

Code : 33

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12706085

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

Parter Medical's Drosophila bottles are available in transparent, autoclavable polypropylene and non-autoclavable polyethylene. The bottle openings are designed to accept standard paper lids, foam plugs, cotton balls and Buzz Plugs.

Round-Bottom Bottles

  • Polypropylene
  • Single use or autoclave for reuse
Square-Bottom Bottles
  • Available in polypropylene or polyethylene
  • Polypropylene provides good transparency and can be autoclaved for reuse or disposed of after single use

Specifications

Specifications

Yes
177mL
Straight wall design minimizes the dislodging of food, provides more efficient racking, filling and plugging
57mm
57mm
Square
Transparent
103mm
Polypropylene
Stock Bottle
Videos
View accessory products