missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Snap-Cap™ Flat-Top Graduated Microcentrifuge Tubes

Available as sterile and nonsterile

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.44000kg

 View more versions of this product

Product Code. 11990069

  • £15.92 / Pack of 500


In Stock 7
Add to basket

Description

Description

 

  • Polypropylene
  • Graduated
  • In different colors

 

Order Info

Sterile tubes are individually wrapped to maintain sterilty

Specifications

Specifications

Snap Cap with Flat Top
1.5mL
14mm
Polypropylene
No
Bulk
Flat Snap
Yellow
39mm
17000xG
0.5mL
Videos