missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Snap-Cap™ Flat-Top Graduated Microcentrifuge Tubes

Available as sterile and nonsterile

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 0.48316kg

 View more versions of this product

Product Code. 12360333

  • £15.92 / Pack of 500


In Stock 1
Add to basket

Specifications

Specifications

Snap Cap with Flat Top
1.5mL
14mm
Polypropylene
No
Bulk
Flat Snap
Blue
39mm
17000xG
0.5mL
Videos