missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Snap-Cap™ Flat-Top Graduated Microcentrifuge Tubes

Available as sterile and nonsterile

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.50760kg

 View more versions of this product

Product Code. 12004897

  • £27.20 / Pack of 1000


In Stock 7
Add to basket

Description

Description

 

  • Polypropylene
  • Graduated
  • In different colors

 

Specifications

Specifications

Snap Cap with Flat Top
0.6mL
10mm
Polypropylene
No
Bulk
Snap
Green
30mm
30000xG
0.2mL
Videos