missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles
Click to view available options
Capacity (Metric):
500 mL
Closure Color:
Assorted
Blue
Natural
Red
Yellow
For Use With (Application):
Acetone
Assorted (Acetone, Distilled Water, Ethanol, Isopropanol, Methanol, Sodium Hypochlorite)
Distilled Water
Ethanol
Isopropanol
Sodium Hypochlorite
Description
Eliminates drips and allows bottle to dispense upright or inverted. Fisherbrand™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles' color-coded polypropylene closures aid in quick and easy identification.
- Valve allows dispensing of wash bottle in an upright or inverted position
- Wide mouth 53mm (2 in.) bottle for easy refilling
- Translucent LDPE bottles with PP valves; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
Compliance
Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Specifications
Capacity | 16 oz., 500mL |
Closure Material | Polypropylene |
Closure Size | 53 mm |
Closure Type | Polypropylene Closure |
Through the Closure Dispensing | Yes |
Product Type | Wash Bottle |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Blue, Natural |
For Use With (Application) | Distilled Water |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'