missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Safety-Labeled Right-to-Know Wash Bottles

Keep liquids in clearly marked bottles that help prevent accidental misuse. Bel Art™ Safety-Labeled Right-to-Know Wash Bottles are available in both preprinted and “Label Your Own” bottles.

Brand:  Fisherbrand™

UNSPSC : 41103209

Code : 41

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12389185

  • £25.11 / Pack of 3


Call for latest availability
+44 (0)1509 555 500
Add to basket

You may only order up to the available stock. For more details call customer services on 01509 555 500 Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 

Specifications

Description

  • Chemical name and formula
  • Full color NFPA™ Diamond
  • Health hazards
  • CAS # (Chemical Abstract Services #)
  • Suggested protective clothing and equipment
  • Color caps enhance identification
  • LDPE bottles with polypropylene caps
  • “Label Your Own” bottles also available
  • Sodium Hypochlorite (Bleach) labeled bottles are white for light protection
  • Use only with solvents compatible with LDPE; Sodium Hypochlorite (Bleach) labeled bottles are WHITE for light protection
Description & Specifications

Specifications

32 oz., 1000mL
OSHA 29 CFR 1910.1200
Yellow
LDPE
No
Wide
1000mL
Polypropylene
Isopropanol
4/Pk.
Yellow
Documentation