missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Rubber-Lined Plastic Caps

Catalog Number 11728709
Click to view available options
Neck Finish:
22-400
38-400
58-400
This item is not returnable. View return policy
This item is not returnable. View return policy

Resilient, resistant to moisture vapor

  • Black, rubber-lined caps Autoclavable
  • Packaged in 12 packs

Closure Liner 14B Rubber
Closure Material Plastic
Closure Size 22-400 mm
Closure Type Screw Cap
For Use With (Equipment) Bottle
Color Black
Description Screw cap
Material Plastic
Length (Metric) 80 mm
No. per Case 576 Cs.
Quantity 576/PK.
Liner Material Rubber
Neck Finish 22-400
Certifications/Compliance ISO 9001
Autoclavable Autoclavable
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'