Learn More
Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks
Manufactured from borosilicate glass. Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks have a sized 10 stopper.
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 2.02500kg
Description
- Borosilicate glass
- Wide neck
- Heavy duty
- Graduated
- Stopper size 10