Learn More
Regenerated Cellulose Dialysis Tubing
Diffuses low molecular weight compounds in aqueous solutions. Fisherbrand™ Regenerated Cellulose Dialysis Tubing retains proteins and bacteria in tubing.
Brand: Fisherbrand
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- Made from regenerated cellulose membrane
- MWCOs from 3500 to 14,000 Daltons
- Individual lot data sheets from outside laboratory supplied with tubing Types T1, T2, and T3; standard grade Type T4 is not tested in outside laboratory
Specifications
Diffuses low molecular weight compounds in aqueous solutions, retains proteins and bacteria in tubing | |
T3 | |
Regenerated Cellulose | |
Two locking nylon membrane clamps, pair of disposable scissors, individual lot data sheets supplied with tubing types T1, T2, and T3, Two locking nylon membrane clamps, pair of disposable surgical scissors, individual lot data sheets supplied with tubing types T1, T2, and T3 | |
Dialysis Tubing |
15m | |
15.9mm | |
Each | |
12000 to 14000 d |