Learn More
Fisherbrand™ Regenerated Cellulose Dialysis Tubing
Diffuses low molecular weight compounds in aqueous solutions. Fisherbrand™ Regenerated Cellulose Dialysis Tubing retains proteins and bacteria in tubing.
Brand:  Fisherbrand™
Code : NEW
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15273847
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • Made from regenerated cellulose membrane
  • MWCOs from 3500 to 14,000 Daltons
  • Individual lot data sheets from outside laboratory supplied with tubing Types T1, T2, and T3; standard grade Type T4 is not tested in outside laboratory
Specifications
Specifications
Diffuses low molecular weight compounds in aqueous solutions, retains proteins and bacteria in tubing
T4
Regenerated Cellulose
Two locking nylon membrane clamps, pair of disposable scissors, individual lot data sheets supplied with tubing types T1, T2, and T3, Two locking nylon membrane clamps, pair of disposable surgical scissors, individual lot data sheets supplied with tubing types T1, T2, and T3
Dialysis Tubing
30m
15.9mm
Each
12000 to 14000 d
Videos