missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Regenerated Cellulose Dialysis Tubing

Diffuses low molecular weight compounds in aqueous solutions. Fisherbrand™ Regenerated Cellulose Dialysis Tubing retains proteins and bacteria in tubing.

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15263857

  • £197.22 / 1 roll


In Stock 4
Add to basket

You may only order up to the available stock. For more details call customer services on 01509 555 500

Description

Description

  • Made from regenerated cellulose membrane
  • MWCOs from 3500 to 14,000 Daltons
  • Individual lot data sheets from outside laboratory supplied with tubing Types T1, T2, and T3; standard grade Type T4 is not tested in outside laboratory

Specifications

Specifications

Diffuses low molecular weight compounds in aqueous solutions, retains proteins and bacteria in tubing
T1
Regenerated Cellulose
Two locking nylon membrane clamps, pair of disposable scissors, individual lot data sheets supplied with tubing types T1, T2, and T3
Dialysis Tubing
15m
29.3mm
Each
3500 d
Videos