Fisherbrand™ Regenerated Cellulose Dialysis Tubing

Diffuses low molecular weight compounds in aqueous solutions; retains proteins and bacteria in tubing

Overview
Brand: Fisherbrand™

DIALYSIS TUBING, 28.6MM DIA, 15M ROLL,12-14KD MWCO

UNSPSC: 41120000

Code: NEW

Additional Details:
Additional Details: Weight: 0.50000kg



Product Code. 15253857

Quantity Price
1 £ 206.57 / 1 roll
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Specification

Item Description Dialysis Tubing
Product Type T3
Kit Contents Two locking nylon membrane clamps, pair of disposable surgical scissors, individual lot data sheets supplied with tubing types T1, T2, and T3
Diameter (Metric) 28.6mm
Material Regenerated Cellulose
Quantity Each
For Use With (Application) Diffuses low molecular weight compounds in aqueous solutions, retains proteins and bacteria in tubing
MWCO 12000 to 14000 d
Length (Metric) 15m