Learn More
Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes provide optically excellent transparency.
Brand: Hellma™ 100-1-40
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Quartz - high UV transmission and low thermal expansion
- Glass - used in the visible range of the spectrum
- Low values of refractive index and dispertion
- Good resistance to chemicals
Specifications
Glass, Quartz, Silica |