Learn More
Hellma Suprasil™ Quartz Micro Cell Cuvettes
Designed to hold samples for spectroscopic experiments and optical analysis. Suprasil™ Quartz Micro Cell Cuvettes are supplied with a PTFE cover.
Brand: Hellma 104-002-10-40
Code : A0
Additional Details : Weight : 0.04950kg
Description
- Sold in pairs
- Very chemically resistant
- Maintains shape, even at high temperatures up to approximately 1000°C
- 80% transmission from 320 to 2500nm
Specifications
Quartz |