Learn More
Fisherbrand™ Polypropylene Rectangular Carboys
Value your lab can rely on. Fisherbrand Polypropylene Rectangular Carboys have a space-saving rectangular design is ideal for storing and dispensing lab water and reagents. Built to last. Made from high quality plastic materials. Autoclavable.
Brand:  Fisherbrand™
UNSPSC : 24111811
Code : 46
Additional Details : Weight : 2.06000kg
 View more versions of this product
Product Code. 13468029
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
5.28 gal., 20L
Yes
Carboy without spigot
Polypropylene
FDA, USP VI
Ideal for storing and dispensing lab water and reagents
Rectangular
20L
Yes
Documentation Alternative Products