missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Graduated Specimen Containers

Ideal for specimen collection, transportation and storage

Brand:  Fisherbrand™

157.23 GBP valid until 2024-12-31
Use promo code "21887" to get your promotional price.



Code : 46

Additional Details : Weight : 7.30000kg

 View more versions of this product

Product Code. 11984375

  • £185.00 / Pack of 500
Estimated Shipment: 15-10-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Autoclavable
  • Graduated
  • With or without nonautoclavable polyethylene snap lid

Specifications

Specifications

No
Polypropylene
8 oz.
Yes
Non-sterile
237 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers