missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polymethylpentene Graduated Cylinders

Calibrated “To Contain/To Deliver” at 20°C

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 1.34000kg

 View more versions of this product

Product Code. 11917884

  • £59.80 / Pack of 8
In Stock
to see stock.
Add to basket

Description

Description

Calibrated “To Contain/To Deliver” at 20°C

  • Transparent cylinders with large molded-in graduations
  • Easy to read
  • Large rounded bases ribbed for reinforcement and stability
  • Generous pour spout; 10mL size has a conical top for easier filling and pouring
  • Non-wetting
  • Chemical, heat and impact resistant
  • Window-clear

Specifications

Specifications

500 mL
To Contain, To Deliver
Autoclavable
Circular
PMP
5 mL
5mL
Graduated Cylinder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers