missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Multipurpose Specimen Storage Containers

Inert to most chemicals including formaldehyde, weak acids and all bases

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 3.44000kg

 View more versions of this product

Product Code. 12089977

  • £41.10 / Pack of 100


Call for latest availability
+44 (0)1509 555 500
Add to basket

For laboratory use only

Description

Description

  • Resist freezing and boiling
  • Stackable for compact storage
  • Select containers are autoclavable (see ordering table); lids are not autoclavable

Specifications

Specifications

High Density Polyethylene
473mL
9.31cm
16.0 oz., 473mL
Low-density polyethylene (not autoclavable)
Translucent
10.81cm
Specimen Container
Videos
Documentation