missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Fisherbrand™ Multipurpose Specimen Storage Containers

Inert to most chemicals including formaldehyde, weak acids and all bases

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 4.90000kg

 View more versions of this product

Product Code. 12009967

  • £56.30 / Pack of 50


Call for latest availability
+44 (0)1509 555 500
Add to basket

For laboratory use only

Description

Description

  • Resist freezing and boiling
  • Stackable for compact storage
  • Select containers are autoclavable (see ordering table); lids are not autoclavable

Specifications

Specifications

High Density Polyethylene
1900mL
14.74cm
64.0 oz., 1900mL
Low-density polyethylene (not autoclavable)
White
15.58cm
Specimen Container
Videos
Documentation