Learn More
Fisherbrand™ Industrial-Grade Vinyl Cleanroom Tape
For light industrial use, clean construction and cleanroom support
Brand:  Fisherbrand™
Code : 03
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13128615
  • £11.00 / Each
Estimated Shipment
14-05-2018
Add to basket
Description
Description
  • Designed for use in areas where ultraclean tape is not required
  • Forms a strong bond with plastics, polymers and stainless steel without leaving adhesive residue when removed
  • Can be written on with cleanroom pen
  • Processed in a controlled environment
  • Wound on a plastic core and sleeve bagged
  • All rolls are 36 yd. long (33m)
Specifications
Specifications
Dark Green
Industrial
33m
Vinyl
Each
FDA, CID
Aisle marking, labeling, batching, clean-build projects, light industrial use and food processing applications
Tape
Industrial grade cleanroom Tape
No
Videos