Fisherbrand™ Industrial-Grade Vinyl Cleanroom Tape

For light industrial use, clean construction and cleanroom support

Overview
Brand: Fisherbrand™

UNSPSC: 41122702

Code: 03

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 13118615

Quantity Price
1 £ 10.65 / Each
Estimated Shipment
12-05-2017
Add to basket


Description and Specification

Specification

Format Tape
Product Type Industrial grade cleanroom Tape
Quantity Each
Sterile No
Description Industrial
Material Vinyl
Length (Metric) 33m
For Use With (Application) Aisle marking, labeling, batching, clean-build projects, light industrial use and food processing applications
Certifications/Compliance FDA, CID
Color Dark Blue