Learn More
Fisherbrand™ Histosette™ II Tissue Processing/Embedding Cassettes with Combination Lid and Base
Fisherbrand™ Histosette™ II Tissue Processing/Embedding Cassettes with Combination Lid and Base open and close as often as necessary — cassettes will always relock securely.
Brand:  Fisherbrand™
Code : 04
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10364511
  • £163.00 / Pack of 1500
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Disposable, plastic cassettes hold tissue specimens during embedding, processing, and storage
  • Molded from a special high-density polymer which is resistant to the chemical action of histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • The anterior writing surface is slanted at a 45° angle, making the cassette more suitable for use with certain types of labeling instruments
Order Info
Available in 11 noncytotoxic, nonmetallic colors.
Specifications
Specifications
Slots
45°
Peach
Tissue Cassette
Tissue Cassette
Combination Lid
Plastic
Videos