missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Heavy-Duty/Utility Funnels

Ideal for continuous lab work and heavy-duty jobs

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.39480kg

 View more versions of this product

Product Code. 12692435

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Yes
210mL
PP (Polypropylene)
12.5cm
14.1cm
Videos
Documentation