missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Heavy-Duty/Utility Funnels

Ideal for continuous lab work and heavy-duty jobs

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 1.34000kg

 View more versions of this product

Product Code. 12682435

  • £33.10 / Pack of 12


In Stock 2
Add to basket

Specifications

Specifications

Yes
100mL
PP (Polypropylene)
8.4cm
11cm
Videos
Documentation