missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Graduated Valumetric™ Bottles without Caps

Store a variety of liquids and solids—dry powders, specimens and soil samples

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 20.44000kg

 View more versions of this product

Product Code. 11533752

  • £137.69 / Pack of 432


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

1 oz., 30mL
Clear
432/PK.
33-400
Screw
30mL
Soda Lime Glass
No
Wide
Videos
Documentation