missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Fluorescence Rectangular Cells: Micro Cells With PTFE Lids
Fit standard spectrophotometer cell holders
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- All cells are 12.5mm wide
- 1, 5 or 10mm path length rectangular cells
- Reduce reagent use with micro cells
- Wavelength range for all cells is 200 to 2500nm
- Quartz windows for transmission from far ultraviolet through the infrared
- Center H: 8.5mm
Specifications
Micro Fluorescence Cell | |
100 μL | |
PTFE | |
2 Ea./Cs. | |
200 to 2500 nm | |
Standard spectrophotometer cell holder | |
Transmission from far ultraviolet through the infrared |
Center H: 8.5mm | |
Quartz | |
2mm | |
10 mm | |
45 mm | |
12.5 mm | |
Rectangle |