missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable Flint Glass Tubes with Plain End

Specially annealed to reduce breakage along stress point

Brand:  Fisherbrand™

Code : 40

Additional Details : Weight : 3.68000kg

 View more versions of this product

Product Code. 15185134

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Round bottom
  • Smooth top and bottom to prevent cuts

Specifications

Specifications

20mm
Specially annealed to reduce breakage along stress point
Tray
36mL
150mm
Disposable Culture Tube
No
Yes
None
250/Tray
KIM-KAP
Flint Glass
Round Bottom
Videos