missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable Flint Glass Tubes with Plain End

Specially annealed to reduce breakage along stress point

Brand:  Fisherbrand™

Code : 40

Additional Details : Weight : 14.34145kg

 View more versions of this product

Product Code. 15195282

  • £97.79 / Pack of 1000


In Stock 448
Add to basket

You may only order up to the available stock. For more details call customer services on 01509 555 500

Description

Description

  • Round bottom
  • Smooth top and bottom to prevent cuts

Specifications

Specifications

16mm
Specially annealed to reduce breakage along stress point
Tray
23mL
150mm
Disposable Culture Tube
No
Yes
None
250/Tray
KIM-KAP
Flint Glass
Round Bottom
Videos