missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Custom Cleaned Clear Boston Round Bottles

Ideal storage for solvents, chemicals and samples. Fisherbrand™ Custom Cleaned Clear Boston Round Bottles are manufactured from clear soda glass.

Brand:  Fisherbrand™

UNSPSC : 24122003

Code : 46

Additional Details : Weight : 4.54000kg

 View more versions of this product

Product Code. 13157474

  • £48.68 / Pack of 12


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Narrow mouth
  • Pre-cleaned for visible particulates using deionised water
  • With white polypropylene cap with PTFE faced foamed polyethylene liner
Specifications

Specifications

16.9 oz., 500mL
USP purified Water
Polyethylene
Soda Lime Glass
Bottle, Boston Round
White
PTFE
Round
500mL
PTFE
Clear
12/PK.
Yes
28-400
Narrow
screw cap
Documentation